Silver foil, 0.025mm (0.001in) thick, annealed, Premion™, 99.998% (metals basis)
Silver foil, 0.025mm (0.001in) thick, annealed, Premion™, 99.998% (metals basis)
Silver foil, 0.025mm (0.001in) thick, annealed, Premion™, 99.998% (metals basis)
Thermo Scientific Chemicals

Silver foil, 0.025mm (0.001in) thick, annealed, Premion™, 99.998% (metals basis)

Form: approx. 0.16g/25x25mm; Temper: soft | CAS: 7440-22-4 | Ag | 107.87 g/mol
Quantity:
25 x 25 mm
50 x 50 mm
50 x 250 mm
Catalog number 042472.GB
also known as 042472-GB
Price (JPY)
-
Quantity:
50 x 250 mm
Request bulk or custom format
Specifications
AppearanceSilver-colored foil
Assay Percent Range99.998% (Metals basis)
CAS7440-22-4
ChEBICHEBI:9141
Chemical Name or MaterialSilver
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only