Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 237
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 237
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 237
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 237
Thermo Scientific Chemicals

Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 237

CAS: 9016-00-6 | (C2H6OSi)n | 74.15 g/mol
Quantity:
50 mL
250 mL
1 L
Catalog number 042496.AK
also known as 042496-AK
Price (JPY)
-
Request A Quote
Quantity:
250 mL
Request bulk or custom format
Chemical Identifiers
CAS9016-00-6
Molecular Formula(C2H6OSi)n
SMILESC[Si](C)(-*)O-*
MDL NumberMFCD00084411
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
Viscosity0.9-1.2 cSt
FormLiquid

Polydimethylsiloxane, trimethylsiloxy is used in water-repellent, protective creams & lotions. It is also used as surfactant, is used in coating auxiliary agents. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only