3-Hydroxy-3-methylbutyric acid, 96%
3-Hydroxy-3-methylbutyric acid, 96%
3-Hydroxy-3-methylbutyric acid, 96%
3-Hydroxy-3-methylbutyric acid, 96%
Thermo Scientific Chemicals

3-Hydroxy-3-methylbutyric acid, 96%

CAS: 625-08-1 | C5H10O3 | 118.13 g/mol
Quantity:
10 g
50 g
250 g
Catalog number 042722.09
also known as 042722-09
Price (JPY)
-
Request A Quote
Quantity:
10 g
Request bulk or custom format
Specifications
Beilstein1743952
CAS625-08-1
ChEBICHEBI:37084
Chemical Name or Material3-Hydroxy-3-methylbutyric acid
ColorYellow
View more
3-Hydroxy-3-methylbutyric acid is a useful biochemical for synthesis and proteomics research. It is used as an indicator of biotin deficiency. Further, it is used to increase muscle mass and decrease muscle breakdown. In addition to this, it is involved in the protein synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only