Thermo Scientific Chemicals

Borane-dimethyl sulfide complex, 2M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 042965.AP
500 mL, Each
Thermo Scientific Chemicals

Borane-dimethyl sulfide complex, 2M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 042965.AP
500 mL, Each
Quantity
Catalog number: 042965.AP
also known as 042965-AP
Price (JPY)

Chemical Identifiers

CAS
13292-87-0
IUPAC Name
(methylsulfanyl)methane borane
Molecular Formula
C2H9BS
InChI Key
RMHDLBZYPISZOI-UHFFFAOYSA-N
SMILES
B.CSC
Appearance (Color)
Clear colorless to pale yellow
Assay (unspecified)
1.90 to 2.20M
Form
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text