Chromium pieces, 1-25mm (0.04-1in), 99.99% (metals basis)
Chromium pieces, 1-25mm (0.04-1in), 99.99% (metals basis)
Chromium pieces, 1-25mm (0.04-1in), 99.99% (metals basis)
Thermo Scientific Chemicals

Chromium pieces, 1-25mm (0.04-1in), 99.99% (metals basis)

CAS: 7440-47-3 | Cr | 52.00 g/mol
Quantity:
50 g
250 g
1 kg
Catalog number 043066.A1
also known as 043066-A1
Price (JPY)
-
Quantity:
1 kg
Request bulk or custom format
Specifications
Assay Percent Notes(metals basis)
CAS7440-47-3
ChEBICHEBI:28073
Chemical Name or MaterialChromium pieces
EINECS Number231-157-5
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only