Thermo Scientific Chemicals

Sodium iodide, ultra dry, 99.98% (metals basis), Thermo Scientific Chemicals

Catalog number: 043447.22
100 g, Each
Thermo Scientific Chemicals

Sodium iodide, ultra dry, 99.98% (metals basis), Thermo Scientific Chemicals

Catalog number: 043447.22
100 g, Each
Quantity
Catalog number: 043447.22
also known as 043447-22
Price (USD)
Price: 411.00
Online price: 349.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7681-82-5
IUPAC Name
sodium iodide
Molecular Formula
INa
InChI Key
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
SMILES
[Na+].[I-]
Appearance (Color)
White
Form
Beads
Total Metal Impurities
≤0.02%

Description

It is used as iodine supplement, expectorant or in its radioactive (I-131) form as an diagnostic aid, particularly for thyroid function tests. sodium Iodide are used in photography and as analytical reagents. It is also used in the measurement of the energy of gamma rays, also used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text