Chloroform, HPLC Grade, 99.5+% min, stab. with amylene
Chloroform, HPLC Grade, 99.5+% min, stab. with amylene
Chloroform, HPLC Grade, 99.5+% min, stab. with amylene
Thermo Scientific Chemicals

Chloroform, HPLC Grade, 99.5+% min, stab. with amylene

CAS: 67-66-3 | CHCl3 | 119.37 g/mol
Quantity:
1 L
4 L
4 x 1 L
4 x 4 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 043685.K2
also known as 043685-K2
Price (JPY)
-
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS67-66-3
IUPAC Nametrichloromethane
Molecular FormulaCHCl3
InChI KeyHEDRZPFGACZZDS-UHFFFAOYSA-N
SMILESClC(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Absorbance at245nm: 1.00AU, max
CommentSubmicron filtered
Impurity contentAcetone and aldehyde ( as acetone): Passes Test
Optical Absorbance at270nm: 0.02AU, max
Optical Absorbance at290nm: 0.01AU, max
View more
Chloroform is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only