Thermo Scientific Chemicals

Cerium(IV) oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 043818.18
50 g, Each
Thermo Scientific Chemicals

Cerium(IV) oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 043818.18
50 g, Each
Quantity
Catalog number: 043818.18
also known as 043818-18
Price (USD)
Price: 219.00
Online price: 189.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1306-38-3
IUPAC Name
dioxocerium
Molecular Formula
CeO2
InChI Key
CETPSERCERDGAM-UHFFFAOYSA-N
SMILES
O=[Ce]=O
Total Metal Impurities
0.1% max.

Description

Cerium oxide is used as a raw material for the production of glass and ceramics as well as additive in paints and varnishes   It is also used in thermal spraying for welding and soldering products and for metal surface treatment products, in industrial polishing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text