Magnesium oxalate dihydrate, 98.5+%
Magnesium oxalate dihydrate, 98.5+%
Magnesium oxalate dihydrate, 98.5+%
Magnesium oxalate dihydrate, 98.5+%
Thermo Scientific Chemicals

Magnesium oxalate dihydrate, 98.5+%

CAS: 547-66-0 | C2H2MgO4+2 | 114.339 g/mol
Quantity:
100 g
500 g
2 kg
Catalog number 043976.22
also known as 043976-22
Price (JPY)
-
Request A Quote
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS547-66-0
IUPAC Namemagnesium(2+) oxalate
Molecular FormulaC2MgO4
InChI KeyUHNWOJJPXCYKCG-UHFFFAOYSA-L
SMILES[Mg++].[O-]C(=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.5% min
Magnesium oxalate dihydrate is in the synthesis of nano sized magnesium oxide. It is also used as a catalyst, refractory materials, adsorbents, superconductors, and ferroelectric materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only