Thermo Scientific Chemicals

Óxido de magnesio, polvo blanco, 96 % mín., Thermo Scientific Chemicals

Número de catálogo: 044078.36
500 g, Each
Thermo Scientific Chemicals

Óxido de magnesio, polvo blanco, 96 % mín., Thermo Scientific Chemicals

Número de catálogo: 044078.36
500 g, Each
Cantidad
Número de catálogo: 044078.36
también denominado 044078-36
Precio (EUR)
Cantidad
-

Identificadores químicos

CAS
1309-48-4
IUPAC Name
oxomagnesium
Molecular Formula
MgO
InChI Key
CPLXHLVBOLITMK-UHFFFAOYSA-N
SMILES
O=[Mg]
Assay (unspecified)
96% min

Descripción

Magnesium oxide is used as a laboratory reagent and an insulator in industrial cables. It acts as a reference material for calorimetry. It is widely used as a basic refractory material for crucibles, a protective coating in plasma displays, an oxide barrier in spin-tunneling devices, an optical material, a component of tubular construction in heating elements, and also in libraries for preserving books. Magnesium oxide microspheres used in enrichment method of polycyclic aromatic hydrocarbons.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text