Pyrogallol, ACS, 99%
Pyrogallol, ACS, 99%
Pyrogallol, ACS, 99%
Thermo Scientific Chemicals

Pyrogallol, ACS, 99%

CAS: 87-66-1 | C6H6O3 | 126.11 g/mol
Quantity:
10 g
50 g
250 g
Catalog number 044152.18
also known as 044152-18
Price (JPY)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS87-66-1
IUPAC Namebenzene-1,2,3-triol
Molecular FormulaC6H6O3
InChI KeyWQGWDDDVZFFDIG-UHFFFAOYSA-N
SMILESOC1=CC=CC(O)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentFe 0.001%
Impurity contentCl 0.001%
Impurity contentSO₄ 0.005%
Residue on ignition0.005%
Trace Metals(as Pb) 5 ppm
Pyrogallol is used in hair dying and suturing materials dying. It is used as a developing agent in black and white developers, as a metal- complexing agent and is involved in oxygen absorption in gas analysis. It exhibits antiseptic properties. Its alkaline solution is used to absorb oxygen from air, thereby it plays a vital role to calculate the amount of oxygen in air.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only