Cobalt(II) acetate tetrahydrate, ACS, 98.0-102.0%
Cobalt(II) acetate tetrahydrate, ACS, 98.0-102.0%
Cobalt(II) acetate tetrahydrate, ACS, 98.0-102.0%
Thermo Scientific Chemicals

Cobalt(II) acetate tetrahydrate, ACS, 98.0-102.0%

CAS: 6147-53-1 | C4H14CoO8 | 249.081 g/mol
Quantity:
25 g
100 g
500 g
Catalog number 044344.14
also known as 044344-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS6147-53-1
IUPAC Nameλ²-cobalt(2+) diacetate tetrahydrate
Molecular FormulaC4H14CoO8
InChI KeyZBYYWKJVSFHYJL-UHFFFAOYSA-L
SMILESO.O.O.O.[Co++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.0-102.0% C₄H₆CoO₄.4H₂O
Impurity contentNi 0.1%
Impurity contentCa 0.005%
Impurity contentCu 0.002%
Impurity contentK 0.01%
View more
Cobalt(II) acetate tetrahydrate is used as an industrial catalyst to harden paints and varnishes, an active catalyst for oxidation and esterification reactions. It is also used to prepare metal complexes having unusual coordination geometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only