Thermo Scientific Chemicals

Copper(II) acetate, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 044355.14
25 g, Each
Thermo Scientific Chemicals

Copper(II) acetate, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 044355.14
25 g, Each
Quantity
Catalog number: 044355.14
also known as 044355-14
Price (USD)
Price: 343.00
Online price: 291.65
Your price:
Quantity
-

Chemical Identifiers

CAS
142-71-2
Total Metal Impurities
0.001% max.

Description

Copper(II) acetate is used in biochemical applications such as DNA extraction. It is used as a source of copper in inorganic synthesis, an oxidizing agent and catalyst in organic synthesis. It is also used to couple two terminal alkynes to make a 1,3-diyne. It is widely used in pigments, manufacture of paris green, textile dyeing, skin conditioner in cosmetics, antioxidant and stabilizer in food grade polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text