Calcium alpha-D-isosaccharinate, 98% (Assay)
Calcium alpha-D-isosaccharinate, 98% (Assay)
Calcium alpha-D-isosaccharinate, 98% (Assay)
Thermo Scientific Chemicals

Calcium alpha-D-isosaccharinate, 98% (Assay)

CAS: 84324-95-8 | C12H22CaO12
Quantity:
5 g
25 g
Catalog number 044371.06
also known as 044371-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS84324-95-8
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Tan
Purity≥97.5%
m alpha-D-isosaccharinate is used to prevent or treat low blood calcium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only