Polystyrene, atactic
Polystyrene, atactic
Polystyrene, atactic
Polystyrene, atactic
Thermo Scientific Chemicals

Polystyrene, atactic

Used as plastics, include protective packaging, containers, lids, bottles, trays, tumblers, and disposable cutlery. | CAS: 9003-53-6 | (C8H8)n | 104.15 g/mol
Quantity:
10 g
50 g
Catalog number 044416.09
also known as 044416-09
Price (JPY)
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS9003-53-6
Molecular Formula(C8H8)n
SMILES*-CC(-*)C1=CC=CC=C1
MDL NumberMFCD00084450
SpecificationsSpecification SheetSpecification Sheet
FormBeads/Pellets
Infrared spectrumConforms
Molecular Weight125,000-250,000
Polystyrene, atactic is used as plastics, include protective packaging, containers, lids, bottles, trays, tumblers, and disposable cutlery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only