Search
View additional product information for Nichrome etchant, Thermo Scientific Chemicals - FAQs (044585.LV)
5 product FAQs found
Nichrome etchant has an etch rate of 50 Å/sec at 40 degrees C.
Undercutting of the etchant can be reduced by increasing the rate of stirring or agitation.
No, it is a ready-to-use solution. Dilution will only be necessary if you want to reduce the etch rate.
The etch rate of the etchant can be reduced by adding 1 part deionized water to 2 parts etchant. This will reduce the etch rate approximately 50%.
The etch rate of the etchant will be increased by increasing the rate of stirring or agitation.