What is the etch rate of Nickel etchant for evaporated nickel films at 30 degrees C?
Nickel etchant for evaporated nickel films has an etch rate of 25 Å/second at 30 degrees C.
Search
View additional product information for Nickel etchant for evaporated nickel films - FAQs (044586.LV, 044586.R0)
5 product FAQs found