Thermo Scientific Chemicals

Tantalum disilicide, Thermo Scientific Chemicals

Catalog number: 044965.18
50 g, Each
Thermo Scientific Chemicals

Tantalum disilicide, Thermo Scientific Chemicals

Catalog number: 044965.18
50 g, Each
Quantity
Catalog number: 044965.18
also known as 044965-18
Price (USD)
Price: 263.00
Online price: 227.65
Your price:
Quantity
-

Chemical Identifiers

CAS
12039-79-1
IUPAC Name
(silanylidenetantalio)silane
Molecular Formula
H4Si2Ta
InChI Key
JSCUKKCIAVANLH-UHFFFAOYSA-N
SMILES
[SiH2]=[Ta]=[SiH2]
Impurity content
1% max

Description

Tantalum disilicide is used in the preparation of ceramics, which finds application in the thermal protection systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text