Gadolinium powder, -200 mesh, 99.9% (REO)
Gadolinium powder, -200 mesh, 99.9% (REO)
Gadolinium powder, -200 mesh, 99.9% (REO)
Gadolinium powder, -200 mesh, 99.9% (REO)
Thermo Scientific Chemicals

Gadolinium powder, -200 mesh, 99.9% (REO)

CAS: 7440-54-2 | Gd | 157.25 g/mol
Quantity:
5 g
25 g
Catalog number 045915.14
also known as 045915-14
Price (JPY)
-
Request A Quote
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS7440-54-2
IUPAC Namegadolinium
Molecular FormulaGd
InChI KeyUIWYJDYFSGRHKR-UHFFFAOYSA-N
SMILES[Gd]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
RUO – Research Use Only