Thermo Scientific Chemicals

Sodium hexafluorosilicate, 99+%, Thermo Scientific Chemicals

Catalog number: 069106.36
500 g, Each
Thermo Scientific Chemicals

Sodium hexafluorosilicate, 99+%, Thermo Scientific Chemicals

Catalog number: 069106.36
500 g, Each
Quantity

Chemical Identifiers

CAS
16893-85-9
IUPAC Name
disodium hexafluorosilanediuide
Molecular Formula
F6Na2Si
InChI Key
TWGUZEUZLCYTCG-UHFFFAOYSA-N
SMILES
[Na+].[Na+].F[Si--](F)(F)(F)(F)F
Form
Crystals or crystalline powder
Assay (unspecified)
≥99.0%

Description

Sodium hexafluorosilicate is used as a additive for water fluoridation, opal glass raw material and in ore refining. It is also used in the preparation of other fluoride chemicals like sodium fluoride, sodium hexafluoroaluminate, potassium hexafluorosilicate, ammonium fluoride and calcium fluoride. It finds application as flotator, component for wet etching, wood preservative, brazing flux and welding compound. It is an active component of concrete.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text