Thermo Scientific Chemicals

Tin(IV) iodide, 95%, Thermo Scientific Chemicals

Catalog number: 071114.09
10 g, Each
Thermo Scientific Chemicals

Tin(IV) iodide, 95%, Thermo Scientific Chemicals

Catalog number: 071114.09
10 g, Each
Quantity
Catalog number: 071114.09
also known as 071114-09
Price (USD)
Price: 64.40
Online price: 54.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7790-47-8
IUPAC Name
tin(IV) iodide
Molecular Formula
H4I4Sn
InChI Key
ZBCVLXKKLMNLGV-UHFFFAOYSA-N
SMILES
[Sn].I.I.I.I
Form
Powder or crystals or crystalline powder
Assay from Supplier's CofA
≥94.0%
Appearance (Color)
Yellow to orange

Description

Used in chemical research and as a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text