Calcium, AAS standard solution, Specpure™ Ca 1000μg/mL
Calcium, AAS standard solution, Specpure™ Ca 1000μg/mL
Calcium, AAS standard solution, Specpure™ Ca 1000μg/mL
Thermo Scientific Chemicals

Calcium, AAS standard solution, Specpure™ Ca 1000μg/mL

CAS: 471-34-1 | CCaO3 | 100.09 g/mol
Quantity:
100 mL
500 mL
Catalog number 088057.AE
also known as 088057-AE
Price (JPY)
-
Quantity:
100 mL
Request bulk or custom format
Specifications
CAS471-34-1
CAS7732-18-5
ChEBICHEBI:3311
Chemical Name or MaterialCalcium
ColorColorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only