Adipic acid, 99%
Adipic acid, 99%
Adipic acid, 99%
Thermo Scientific Chemicals

Adipic acid, 99%

CAS: 124-04-9 | C6H10O4 | 146.142 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number 102815000
Price (USD)
30.65
Online Exclusive
34.00
Save 3.35 (10%)
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
30.65
Online Exclusive
34.00
Save 3.35 (10%)
Each
Chemical Identifiers
CAS124-04-9
IUPAC Namehexanedioic acid
Molecular FormulaC6H10O4
InChI KeyWNLRTRBMVRJNCN-UHFFFAOYSA-N
SMILESOC(=O)CCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point151°C to 153°C
Titration with NaOH>=99 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only