TRIzol™ LS Reagent
Invitrogen™
TRIzol™ LS Reagent
TRIzol™ LS Reagent is a complete, ready-to-use reagent optimized for the isolation of high-quality total RNA, or the simultaneous isolationRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
10296028200 mL
10296010100 mL
Catalog number 10296028
Price (USD)
550.65
Online exclusive
618.00
Save 67.35
Each
Quantity:
200 mL
Price (USD)
550.65
Online exclusive
618.00
Save 67.35
Each
TRIzol™ LS Reagent is a complete, ready-to-use reagent optimized for the isolation of high-quality total RNA, or the simultaneous isolation of RNA, DNA, and protein from a variety of liquid samples. This monophasic solution of phenol and guanidine isothiocyanate is designed to isolate separate fractions of RNA, DNA, and proteins from liquid samples of human, animal, plant, yeast, bacterial, and viral origin, typically within one hour.
Key features of TRIzol™ LS Reagent:
• Formulated for use with liquid samples such as serum and virus preparations
• Facilitates recovery of RNA, DNA, and protein from a single liquid sample
• Offers excellent lysis capability, even with difficult biological fluids
Reliably purify RNA from multiple sample sources
TRIzol™ LS Reagent is designed for processing a variety of liquid samples of up to 0.25 mL in volume. TRIzol™ LS Reagent differs from the standard TRIzol™ Reagent in concentration, which permits larger samples to be processed. Just as with the standard TRIzol™ Reagent, the integrity of resulting RNA preparations is maintained by the highly effective inhibition of RNase activity during sample homogenization. The simplicity of the TRIzol™ LS Reagent method allows simultaneous processing of a large number of samples. The entire procedure can be completed in 1 hour. Total RNA isolated by TRIzol™ LS Reagent is free of protein and DNA contamination.
Formulated to serve multiple isolations
TRIzol™ LS Reagent allows you to perform sequential precipitation of RNA, DNA, and proteins from a single sample. After homogenizing the sample with TRIzol™ LS Reagent, chloroform is added, and the homogenate is allowed to separate into a clear upper aqueous layer (containing RNA), an interface, and a red lower organic layer (containing the DNA and proteins). RNA is precipitated from the aqueous layer with isopropanol. DNA is precipitated from the aqueous/organic interface with ethanol. Protein is precipitated from the phenol-ethanol layer by isopropanol precipitation. The precipitated RNA, DNA, or protein is washed to remove impurities, and then resuspended for use in downstream applications.
Purified products are ideal for use with a variety of applications
Isolated RNA can be used in real-time quantitative PCR (qPCR), northern blot analysis, dot blot hybridization, poly-(A)+ selection, In vitro translation, RNase protection assays, and molecular cloning. Isolated DNA can be used in PCR, restriction enzyme digestion, and Southern blots. Isolated protein can be used for western blots, recovery of some enzymatic activity, and some immunoprecipitation
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Elution Volume20 to 600 μL
Final Product TypeTotal RNA, DNA, Protein
For Use With (Application)RT-PCR, qPCR, cDNA library construction, NGS, microarray analysis, blot hybridization, Northern/Southern/Western blotting, in vitro translation, nuclease protection assays, nucleic acid labeling, hybridization, enzymatic assays, immunoprecipitation, gel shift assays, 2D gel electrophoresis
High-throughput CompatibilityNot High-throughput Compatible (Manual)
Purification Time1 hr.
Quantity200 mL
Shipping ConditionRoom Temperature
Starting Material Amount0.25 mL per 0.75 mL of TRIzol™ LS Reagent
YieldRNA: ≤20 μg per 1 mL blood
≤1.4 μg per 106 cells
Isolation TechnologyOrganic Extraction
Sample TypeLiquid Samples (e.g. Serum)
Unit SizeEach
Contents & Storage
200 mL TRIzol LS Reagent; room temperature
Frequently asked questions (FAQs)
What chloroform do you recommend I use for RNA extraction with TRIzol Reagent? Are there any substitutes I can use?
We recommend using straight chloroform. No isoamyl alcohol is needed (though using chloroform:isoamyl alcohol 49:1 works without problems). You can also use chloroform with 50 ppm amylene. Alternatively, BCP (1-bromo-2 chloropropane) can be used in the place of chloroform.
Find additional tips, troubleshooting help, and resources within our RNA Sample Collection, Protection, and Isolation Support Center.
What is the smallest sample volume I can use when extracting RNA with TRIzol Reagent?
Small volumes (0.5-0.8 mL) of TRIzol Reagent have been used successfully for 10^2 to 10^5 cells, but if small volumes are used, we recommend using smaller tubes in order to have the tallest possible column of aqueous phase. The taller the column of liquid, the less likely that contamination from the interphase will occur.
Here is a protocol for isolation of RNA from small quantities of tissue (1-10 mg) or cells (100-10,000):
1. Add 800 µL TRIzol Reagent to the sample. Homogenize cells by pipetting repeatedly. Add 200 µg glycogen (Cat. No. 10814010) directly to the TRIzol Reagent. If processing tissue, pulverize in liquid nitrogen first and then add 800 µL TRIzol Reagent containing 200 µg glycogen (final concentration 250 µg/mL) followed by vigorous vortexing or power homogenization.
2. Place at room temperature, cap the vial, and vortex at high speed for 10 seconds. Make sure the TRIzol Reagent wets the side of the vial in order to solubilize any sample that may be remaining on the walls.
3. Shear the genomic DNA in the sample by passing twice through a 26-gauge needle connected to a 1 mL syringe. Using the syringe, transfer the sample to a sterile 1.5 mL microcentrifuge tube.
4. Add 160 µL of chloroform (or 49:1 chloroform:isoamyl alcohol) to each sample and vortex up to 30 seconds. Centrifuge at maximum speed in a microcentrifuge for 5 minutes to separate the phases.
5. Transfer the upper aqueous phase to a fresh tube and add 400 µL ice-cold isopropanol. Allow the samples to precipitate at -20 degrees C for 1 hour to overnight. Pellet the RNA by centrifugation at maximum speed in the microfuge for 15 minutes at room temperature.
6. Decant the supernatant. Wash the pellet in 200 µL of 70% ethanol and centrifuge again for 10 minutes at maximum speed. Decant the supernatant, removing as much as possible without disturbing the pellet. Dry the RNA pellet.
7. Resolubilize the pellet in 30-50 µL RNAse-free deionized water. If tissue is high in RNAses (e.g., adrenal gland, pancreas), resuspend in 100% deionized formamide. Be sure to vortex or pipette the sample up and down to ensure that the pellet is fully resolubilized. Store at -70 degrees C.
Find additional tips, troubleshooting help, and resources within ourRNA Sample Collection, Protection, and Isolation Support Center.
What are possible stopping points and storage for RNA extraction when using TRIzol Reagent for RNA extraction? How should I store the RNA?
There are several possible stopping points and recommended storage conditions during the extraction of RNA with TRIzol Reagent:
- Sample homogenization step: After homogenization (before addition of chloroform), you can store samples at -70 degrees C for at least 1 year. The homogenated samples can sit at room temperature for several hours before adding chloroform.
- Sample homogenization step: If samples are efficiently lysed in TRIzol Reagent and the reagent inactivates the nucleases, you can safely store RNA for 3-4 days at room temperature.
- RNA precipitation step: You can store RNA in isopropanol overnight at 4 degrees C. Prolonged storage at this reduced temperature will not influence the yield of RNA appreciably. Do not store at -20 degrees C, as salts will precipitate, and do not store for a prolonged time at room temperature because the guanidine isothiocyanate can harm the RNA.
- RNA wash step: You can store RNA in 75% ethanol for 1 week at 4 degrees C or 1 year at -20 degrees C.
Find additional tips, troubleshooting help, and resources within our RNA Sample Collection, Protection, and Isolation Support Center.
What is the excepted A260/A280 absorbance ratio of total RNA isolated by TRIzol Reagent?
The absorbance of nucleic acids is dependent upon the ionic strength and pH of the medium. Please see the range of absorbance values below based on the diluents used.
Diluent A260 A280 A260/A280 RNA (µg/mL)
Cytoplasmic RNA dissolved in distilled water 0.381 0.223 1.711 15.24
Cytoplasmic RNA dissolved in TE buffer 0.335 0.145 2.310 13.4
RNA isolated by TRIzol Reagent and dissolved in distilled water 0.585 0.328 1.785 23.4 RNA isolated by TRIzol Reagent and dissolved in TE buffer 0.544 0.247 2.206 21.76 Although a high A260/A280 ratio may not indicate an extremely pure preparation of nucleic acid, a low A260/A280 ratio (1.7 for RNA) does indicate that the preparation is contaminated and may not be suitable for some applications.
Find additional tips, troubleshooting help, and resources within our RNA Sample Collection, Protection, and Isolation Support Center.
Do you have any tips for RNA isolation when working with blood samples?
Please visit our website for tips for working with blood samples.
Find additional tips, troubleshooting help, and resources within our RNA Sample Collection, Protection, and Isolation Support Center.
Citations & References (5)
Citations & References
Abstract
Authors:
Journal:
PubMed ID:16475517
Human Cytomegalovirus Induces Drug Resistance and Alteration of Programmed Cell Death by Accumulation of Delta N-p73alpha.
Authors: Allart Sophie; Martin Helene; Detraves Claire; Terrasson Jerome; Caput Daniel; Davrinche Christian;
Journal:J Biol Chem
PubMed ID:12034725
'Intrauterine transmission of human cytomegalovirus (HCMV) to the fetus following primary infection in early and late pregnancy usually results in severe neurological handicaps and sensorineural hearing loss with typical migrational anomalies, optic atrophy, disturbed myelination, cerebella hypoplasia, microcephaly, hydrocephaly, and lissencephaly. Recently, evidences raised from the phenotype of p73-deficient mice ... More
Opposing actions of inositol 1,4,5-trisphosphate and ryanodine receptors on nuclear factor of activated T-cells regulation in smooth muscle.
Authors:Gomez MF, Stevenson AS, Bonev AD, Hill-Eubanks DC, Nelson MT,
Journal:J Biol Chem
PubMed ID:12145283
'The nuclear factor of activated T-cells (NFAT), originally identified in T-cells, has since been shown to play a role in mediating Ca(2+)-dependent gene transcription in diverse cell types outside of the immune system. We have previously shown that nuclear accumulation of NFATc3 is induced in ileal smooth muscle by platelet-derived ... More
A virus discovery method incorporating DNase treatment and its application to the identification of two bovine parvovirus species.
Authors: Allander T; Emerson S U; Engle R E; Purcell R H; Bukh J;
Journal:Proc Natl Acad Sci U S A
PubMed ID:11562506
Identification of previously unrecognized viral agents in serum or plasma samples is of great medical interest but remains a major challenge, primarily because of abundant host DNA. The current methods, library screening or representational difference analysis (RDA), are very laborious and require selected sample sets. We have developed a simple ... More
Focal Adhesion Kinase Activated by beta 4 Integrin Ligation to mCLCA1 Mediates Early Metastatic Growth.
Authors: Abdel-Ghany Mossaad; Cheng Hung-Chi; Elble Randolph C; Pauli Bendicht U;
Journal:J Biol Chem
PubMed ID:12110680
Early metastatic growth occurs at sites of vascular arrest of blood-borne cancer cells and is entirely intravascular. Here we show that lung colonization by B16-F10 cells is licensed by beta(4) integrin adhesion to the mouse lung endothelial Ca(2+)-activated chloride channel protein mCLCA1. In a manner independent of Met, beta(4) integrin-mCLCA1-ligation ... More