4-Aminoveratrole, 98%
4-Aminoveratrole, 98%
4-Aminoveratrole, 98%
Thermo Scientific Chemicals

4-Aminoveratrole, 98%

CAS: 6315-89-5 | C8H11NO2 | 153.18 g/mol
Quantity:
10 g
25 g
Packaging:
Glass bottle
Catalog number 104720250
Price (JPY)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6315-89-5
IUPAC Name3,4-dimethoxyaniline
Molecular FormulaC8H11NO2
InChI KeyLGDHZCLREKIGKJ-UHFFFAOYSA-N
SMILESCOC1=CC=C(N)C=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown to purple
Melting point85°C to 89°C
Appearance (Form)Powder
GC>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only