1,2,4,5-Benzenetetracarboxylic anhydride, 99%
1,2,4,5-Benzenetetracarboxylic anhydride, 99%
1,2,4,5-Benzenetetracarboxylic anhydride, 99%
Thermo Scientific Chemicals

1,2,4,5-Benzenetetracarboxylic anhydride, 99%

CAS: 89-32-7 | C10H2O6 | 218.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 105321000
Price (USD)
53.65
Special offer
Online exclusive
Ends: 28-Dec-2026
63.40
Save 9.75
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
53.65
Special offer
Online exclusive
Ends: 28-Dec-2026
63.40
Save 9.75
Each
Chemical Identifiers
CAS89-32-7
IUPAC Name5,11-dioxatricyclo[7.3.0.0³,⁷]dodeca-1,3(7),8-triene-4,6,10,12-tetrone
Molecular FormulaC10H2O6
InChI KeyANSXAPJVJOKRDJ-UHFFFAOYSA-N
SMILESO=C1OC(=O)C2=CC3=C(C=C12)C(=O)OC3=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Appearance (Form)Powder
Infrared spectrumConforms
Melting point283°C to 287°C
Titration excess NaOH98.5 to 101.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only