Benzo[ghi]perylene, 98+%
Benzo[ghi]perylene, 98+%
Benzo[ghi]perylene, 98+%
Acros Organics

Benzo[ghi]perylene, 98+%

CAS: 191-24-2 | C22H12 | 276.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
5 mg
25 mg
Packaging:
Glass bottle
Catalog number 105551000
HSN 29029090
Price (INR)
45,757.00
Each
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
45,757.00
Each
Chemical Identifiers
CAS191-24-2
IUPAC Namehexacyclo[12.8.0.0²,¹¹.0³,⁸.0⁴,²¹.0¹⁷,²²]docosa-1(22),2,4,6,8,10,12,14,16,18,20-undecaene
Molecular FormulaC22H12
InChI KeyGYFAGKUZYNFMBN-UHFFFAOYSA-N
SMILESC1=CC2=CC=C3C=CC4=CC=C5C=CC=C6C(=C1)C2=C3C4=C56
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point276°C to 280°C
Appearance (Form)Crystalline powder or crystals or needles
Appearance (Color)Yellow to orange
Infrared spectrumConforms
GC-MS>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only