sec-Butylamine, 99%
sec-Butylamine, 99%
sec-Butylamine, 99%
Thermo Scientific Chemicals

sec-Butylamine, 99%

CAS: 13952-84-6 | C4H11N | 73.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
107815000500 mLGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 107815000
Price (USD)
-
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS13952-84-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.3920 to 1.3960 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only