n-Butyl disulfide, 97%
n-Butyl disulfide, 97%
n-Butyl disulfide, 97%
Thermo Scientific Chemicals

n-Butyl disulfide, 97%

CAS: 629-45-8 | C8H18S2 | 178.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 107920250
Price (USD)
41.65
Special offer
Online exclusive
Ends: 28-Dec-2026
48.90
Save 7.25
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
41.65
Special offer
Online exclusive
Ends: 28-Dec-2026
48.90
Save 7.25
Each
Chemical Identifiers
CAS629-45-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light brown
GC>=96.0 %
Infrared spectrumConforms
Refractive index1.4915 to 1.4935 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only