Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: YGTMVFVRS FLVGEAARAL SKACTIAIRY SAVRHQSEMK PGEPEPQILD FQTQQYKLFP LLATAYAFQF VGAYMKETYH RINEGIGQGD LSELPELHAL TAGLKAFTSW TANTGIEACR MACGGHGYSH CSGLPNIYVN FTPSCTFEGE NTVMMLQTAR FLMKSYDQVH SGKLVCGMVS YLNDLPSQRI QPQQVAVWPT MVDINSPESL TEAYKLRAAR LVEIAAKNLQ KEVIHRKSKE VAWNLTSVDL VRASEAHCHY VVVKLFSEKL LKIQDKAIQA VLRSLCLLYS LYGISQNAGD FLQGSIMTEP QITQVNQRVK ELLTLIRSDA VALVDAFDFQ DVTLGSVLGR YDGNVYENLF EWAKNSPLNK AEVHESYKHL KSLQSKL (275-660 aa encoded by BC010425 )
The protein encoded by this gene is the first enzyme of the fatty acid beta-oxidation pathway, which catalyzes the desaturation of acyl-CoAs to 2-trans-enoyl-CoAs. It donates electrons directly to molecular oxygen, thereby producing hydrogen peroxide. Defects in this gene result in pseudoneonatal adrenoleukodystrophy, a disease that is characterized by accumulation of very long chain fatty acids. Alternatively spliced transcript variants encoding different isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ACO; acyl-CoA oxidase 1, palmitoyl; acyl-CoA oxidase, straight-chain; acyl-Coenzyme A oxidase 1, palmitoyl; AOX; Palmitoyl-CoA oxidase; Peroxisomal acyl-coenzyme A oxidase 1; Peroxisomal fatty acyl-CoA oxidase; peroxisomal fatty acyl-coenzyme A oxidase; peroxisomal straight-chain Acyl-CoA oxidase; SCOX; Straight-chain acyl-CoA oxidase; unnamed protein product
Gene Aliases: ACOX; ACOX1; AOX; D130055E20Rik; MITCH; PALMCOX; Paox; RATACOA1; SCOX
UniProt ID: (Human) Q15067, (Rat) P07872, (Mouse) Q9R0H0
Entrez Gene ID: (Human) 51, (Rat) 50681, (Mouse) 11430
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support