Thermo Scientific Chemicals

4-Chlorostyrene, 99%, stabilized, Thermo Scientific Chemicals

Catalog number: 110090500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Chlorostyrene, 99%, stabilized, Thermo Scientific Chemicals

Catalog number: 110090500
50 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
1073-67-2
IUPAC Name
1-chloro-4-ethenylbenzene
Molecular Formula
C8H7Cl
InChI Key
KTZVZZJJVJQZHV-UHFFFAOYSA-N
SMILES
ClC1=CC=C(C=C)C=C1
Appearance (Form)
Liquid
GC
>=98.5 %
Stabilizer
0.1 % tert.-butylcatechol
Refractive index
1.5645 to 1.5665 (20°C, 589 nm)
Appearance (Color)
Clear colorless to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text