Nalgene™ Cylindrical Polypropylene Tank with Cover, 7.5 gallon - FAQs
View additional product information for Nalgene™ Cylindrical Polypropylene Tank with Cover - FAQs (11200-0007, 11200-0005, 11200-0015, 11200-0100, 11200-0055, 11200-0010, 11200-0030)
1 product FAQs found
Can I buy a replacement lid for my Nalgene Cylindrical Polypropylene Tank with Cover?
Our Nalgene Cylindrical Polypropylene Tanks are molded as one piece and then cut to create the lid. As a result, we do not have replacement lids.