9,10-Dibromoanthracene, 96%
9,10-Dibromoanthracene, 96%
9,10-Dibromoanthracene, 96%
Thermo Scientific Chemicals

9,10-Dibromoanthracene, 96%

CAS: 523-27-3 | C14H8Br2 | 336.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 112710500
Price (TWD)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS523-27-3
IUPAC Name9,10-dibromoanthracene
Molecular FormulaC14H8Br2
InChI KeyBRUOAURMAFDGLP-UHFFFAOYSA-N
SMILESBrC1=C2C=CC=CC2=C(Br)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to yellow-green
Appearance (Form)Powder
Infrared spectrumConforms
Melting point220°C to 225°C
GC>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only