1,4-Dicyanobenzene, 98%
1,4-Dicyanobenzene, 98%
1,4-Dicyanobenzene, 98%
Thermo Scientific Chemicals

1,4-Dicyanobenzene, 98%

CAS: 623-26-7 | C8H4N2 | 128.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 113825000
Price (EUR)
332,00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
332,00
Each
Chemical Identifiers
CAS623-26-7
IUPAC Namebenzene-1,4-dicarbonitrile
Molecular FormulaC8H4N2
InChI KeyBHXFKXOIODIUJO-UHFFFAOYSA-N
SMILESN#CC1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Appearance (Form)Powder
Appearance (Color)White to light yellow to light beige
Infrared spectrumConforms
Melting point223°C to 230°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only