3,4-Dimethoxybenzoic acid, 99+%
3,4-Dimethoxybenzoic acid, 99+%
3,4-Dimethoxybenzoic acid, 99+%
Thermo Scientific Chemicals

3,4-Dimethoxybenzoic acid, 99+%

CAS: 93-07-2 | C9H10O4 | 182.18 g/mol
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 115450250
Price (JPY)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS93-07-2
IUPAC Name3,4-dimethoxybenzoic acid
Molecular FormulaC9H10O4
InChI KeyDAUAQNGYDSHRET-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1OC)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)White to beige
Infrared spectrumConforms
Melting point180°C to 184°C
HPLC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only