Thermo Scientific Chemicals

3,5-Dimethylbenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: 116000250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

3,5-Dimethylbenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: 116000250
25 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
499-06-9
IUPAC Name
3,5-dimethylbenzoic acid
Molecular Formula
C9H10O2
InChI Key
UMVOQQDNEYOJOK-UHFFFAOYSA-N
SMILES
CC1=CC(=CC(C)=C1)C(O)=O
Infrared spectrum
Conforms
Titration with NaOH
98.5 to 101.5 %
Appearance (Color)
White to yellow
Melting point
169°C to 172°C
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text