Ethanethiol, 99+%
Ethanethiol, 99+%
Ethanethiol, 99+%
Thermo Scientific Chemicals

Ethanethiol, 99+%

CAS: 75-08-1 | C2H6S | 62.13 g/mol
Quantity:
50 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 117860500
Price (JPY)
-
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS75-08-1
IUPAC Nameethanethiol
Molecular FormulaC2H6S
InChI KeyDNJIEGIFACGWOD-UHFFFAOYSA-N
SMILESCCS
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
GC>=99.0 %
Infrared spectrumConforms
Refractive index1.4296 to 1.4316 (20°C, 589 nm)
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only