DL-Ethyl 2-bromopropionate, 99%
DL-Ethyl 2-bromopropionate, 99%
DL-Ethyl 2-bromopropionate, 99%
Thermo Scientific Chemicals

DL-Ethyl 2-bromopropionate, 99%

CAS: 535-11-5 | C5H9BrO2 | 181.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number 118140010
Price (USD)
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS535-11-5
IUPAC Nameethyl 2-bromopropanoate
Molecular FormulaC5H9BrO2
InChI KeyARFLASKVLJTEJD-UHFFFAOYNA-N
SMILESCCOC(=O)C(C)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
GC>=98.5 %
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.4450 to 1.4470 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only