Ethyl isonipecotate, 98+%
Ethyl isonipecotate, 98+%
Ethyl isonipecotate, 98+%
Thermo Scientific Chemicals

Ethyl isonipecotate, 98+%

CAS: 1126-09-6 | C8H15NO2 | 157.21 g/mol
Have Questions?
Catalog NumberQuantityPackaging
11864025025 gGlass bottle
Catalog number 118640250
Price (USD)
45.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
45.00
Each
Chemical Identifiers
CAS1126-09-6
SpecificationsSpecification SheetSpecification Sheet
GC>=98.0 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light brown
Infrared spectrumConforms
Refractive index1.4585 to 1.4605 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only