1-Fluoro-2-nitrobenzene, 99%
1-Fluoro-2-nitrobenzene, 99%
1-Fluoro-2-nitrobenzene, 99%
Thermo Scientific Chemicals

1-Fluoro-2-nitrobenzene, 99%

CAS: 1493-27-2 | C6H4FNO2 | 141.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
Catalog number 119482500
Price (TWD)
-
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1493-27-2
IUPAC Name1-fluoro-2-nitrobenzene
Molecular FormulaC6H4FNO2
InChI KeyPWKNBLFSJAVFAB-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to brown
Infrared spectrumConforms
Refractive index1.5307 to 1.5327 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only