4-Fluorophenylhydrazine hydrochloride, 97%
4-Fluorophenylhydrazine hydrochloride, 97%
4-Fluorophenylhydrazine hydrochloride, 97%
Thermo Scientific Chemicals

4-Fluorophenylhydrazine hydrochloride, 97%

CAS: 823-85-8 | C6H8ClFN2 | 162.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 119590100
Price (USD)
56.65
Special offer
Online exclusive
Ends: 28-Dec-2026
66.80
Save 10.15
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
56.65
Special offer
Online exclusive
Ends: 28-Dec-2026
66.80
Save 10.15
Each
Chemical Identifiers
CAS823-85-8
IUPAC Namehydrogen (4-fluorophenyl)hydrazine chloride
Molecular FormulaC6H8ClFN2
InChI KeyFEKUXLUOKFSMRO-UHFFFAOYSA-N
SMILES[H+].[Cl-].NNC1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=96.0 %
Appearance (Color)White to brown
Infrared spectrumConforms
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only