DMEM, no glucose - FAQs
View additional product information for DMEM, no glucose - FAQs (11966025)
2 product FAQs found
My cells will not grow in DMEM, what other type of culture media can be used with the Photoreactive Amino Acids?
Some cells types can be adapted to grow in DMEM before using the DMEM-LM supplemented with the photoreactive amino acids. Currently we do not offer any other leucine- and methionine-depleted culture medium.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What is the manganese concentration in DMEM? Do you offer manganese-free DMEM?
Manganese is not present in the formulation of our catalog DMEM media products.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.