DMEM, no glucose
DMEM, no glucose
DMEM, no glucose
Gibco™

DMEM, no glucose

DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells.Read more
Have Questions?
Catalog NumberQuantity
11966025500 mL
Catalog number 11966025
Price (MXN)
1,009.69
Each
Quantity:
500 mL
Customize this product
Price (MXN)
1,009.69
Each

DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. We offer a variety of DMEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.


This DMEM is modified as follows:
WithWithout
• L-glutamine• Glucose
• Phenol Red• Sodium Pyruvate
 • HEPES


The complete formulation is available.

Using DMEM
DMEM is unique from other media as it contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate. DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM uses a sodium bicarbonate buffer system (3.7 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell LineHeLa, 293, Cos-7, and PC-12
Cell TypePrimary Fibroblasts, Neurons, Glial Cells, HUVECs, Smooth Muscle Cells
Concentration1 X
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco
Product TypeDMEM (Dulbecco's Modified Eagle Medium)
Quantity500 mL
Shelf Life12 Months From Date of Manufacture
Shipping ConditionRoom Temperature
ClassificationAnimal Origin-free
FormLiquid
Serum LevelStandard Serum Supplementation
SterilitySterile-filtered
With AdditivesGlutamine, Phenol Red
Without AdditivesNo Glucose, No HEPES, No Sodium Pyruvate
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 8°C (protect from light)
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Frequently asked questions (FAQs)

My cells will not grow in DMEM, what other type of culture media can be used with the Photoreactive Amino Acids?

Some cells types can be adapted to grow in DMEM before using the DMEM-LM supplemented with the photoreactive amino acids. Currently we do not offer any other leucine- and methionine-depleted culture medium.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

What is the manganese concentration in DMEM? Do you offer manganese-free DMEM?

Manganese is not present in the formulation of our catalog DMEM media products.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (3)

Citations & References
Abstract
Coupling of glucose deprivation with impaired histone H2B monoubiquitination in tumors.
Authors:Urasaki Y, Heath L, Xu CW
Journal:PLoS One
PubMed ID:22615809
Metabolic reprogramming is associated with tumorigenesis. However, glucose metabolism in tumors is poorly understood. Here, we report that glucose levels are significantly lower in bulk tumor specimens than those in normal tissues of the same tissue origins. We show that mono-ubiquitinated histone H2B (uH2B) is a semi-quantitative histone marker for ... More
The role of p53 deacetylation in p21Waf1 regulation by laminar flow.
Authors:Zeng L, Zhang Y, Chien S, Liu X, Shyy JY,
Journal:J Biol Chem
PubMed ID:12716906
Laminar flow arrests vascular endothelial cells at the G0/G1 phase with concurrent increase in p53 and p21Waf1. We investigated the molecular mechanism by which laminar flow activates p53 and p21Waf1 in endothelial cells. The application of a laminar flow (12 dyn/cm2) increased the deacetylation at Lys-320 and Lys-373 of p53 ... More
Glucose uptake mediated by glucose transporter 1 is essential for early tooth morphogenesis and size determination of murine molars.
Authors:Ida-Yonemochi H, Nakatomi M, Harada H, Takata H, Baba O, Ohshima H
Journal:Dev Biol
PubMed ID:22226978
Glucose is an essential source of energy for body metabolism and is transported into cells by glucose transporters (GLUTs). Well-characterized class I GLUT is subdivided into GLUTs1-4, which are selectively expressed depending on tissue glucose requirements. However, there is no available data on the role of GLUTs during tooth development. ... More