Acide gibbérellique, 90 %, Thermo Scientific Chemicals
Acide gibbérellique, 90 %, Thermo Scientific Chemicals
Acide gibbérellique, 90 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acide gibbérellique, 90 %, Thermo Scientific Chemicals

CAS: 77-06-5 | C19H22O6 | 346.38 g/mol
Have Questions?
Modifier l'affichagebuttonViewtableView
Quantité:
1 g
5 g
25 g
Conditionnement:
Flacon en verre
Référence 119860250
Prix (EUR)
490,00
Each
Quantité:
25 g
Conditionnement:
Flacon en verre
Grand volume ou format personnalisé
Prix (EUR)
490,00
Each
Identifiants chimiques
CAS77-06-5
IUPAC Name(1R,2R,5S,8S,9S,10R,12S)-5,12-dihydroxy-11-methyl-6-methylidene-16-oxo-15-oxapentacyclo[9.3.2.1⁵,⁸.0¹,¹⁰.0²,⁸]heptadec-13-ene-9-carboxylic acid
Molecular FormulaC19H22O6
InChI KeyIXORZMNAPKEEDV-QTWFBFKQSA-N
SMILESCC12[C@H]3[C@H](C(O)=O)[C@@]45CC(=C)[C@@](O)(C4)CC[C@H]5[C@]3(OC1=O)C=C[C@@H]2O
Voir plus
Appearance (Color)White to light yellow
Loss on drying=<3 % (100°C, 7 hrs) (vacuum)
Arsenic (As)=<3 ppm
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Voir plus
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only