Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • RELM alpha Antibodies

          Invitrogen

          RELM alpha Monoclonal Antibody (DS8RELM), PE, eBioscience™

          Advanced Verification
          5 Published Figures
          3 References
          View all (12) RELM alpha antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite RELM alpha Monoclonal Antibody (DS8RELM), PE, eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (5)
          • Advanced Verification (2)
          RELM alpha Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          RELM alpha Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 8

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RELM alpha Antibody (12-5441-80) in Flow

          C57BL/6 mouse resident peritoneal exudate cells were surface stained with F4/80 Monoclonal Antibody, eFluor 450 (Product # 48-4801-82). The cells were then fixed and permeabilized using the Intracellular Fixation & Permeabilization Buffer Set (Product # 88-8824-00) and protocol, and intracellularly stained with either 0.25 µg of Rat IgG1 kappa Isotype Control, PE (Product # 12-4301-82) (left) or 0.25 µg of RELM alpha Monoclonal Antibody, PE (Product # 12-5441-82)(right). All cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RELM alpha Antibody in Flow Cytometry (Flow)
          RELM alpha Antibody in Flow Cytometry (Flow)
          RELM alpha Antibody in Flow Cytometry (Flow)
          RELM alpha Antibody in Flow Cytometry (Flow)
          RELM alpha Antibody in Flow Cytometry (Flow)
          RELM alpha Antibody in Flow Cytometry (Flow)
          RELM alpha Antibody
          RELM alpha Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          12-5441-80

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          0.5 µg/test
          View 2 publications 2 publications

          Functional Assay (Functional)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Mouse

          Host/Isotype

          Rat / IgG1, kappa

          Recommended Isotype Control

          Rat IgG1 kappa Isotype Control (eBRG1), PE, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          DS8RELM

          Immunogen

          Recombinant mouse RELM alpha protein

          Conjugate

          PE PE PE
          • Alexa Fluor 700 
          • APC 
          • Brilliant Violet 421
          • PerCP-eFluor 710
          • Request custom conjugation

          Excitation/Emission Max

          565/576 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          25 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2762681

          Product Specific Information

          Description: This DS8RELM monoclonal antibody recognizes mouse RELM alpha, also known as FIZZ1. This antibody was shown not to cross-react with mouse RELM gamma.

          Applications Reported: This DS8RELM antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This DS8RELM antibody has been tested by flow cytometric analysis of resident peritoneal macrophages using the Intracellular Fixation & Permeabilization Buffer Set (Product # 88-8824-00) and protocol. Please refer to BestProtocols®: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins located under the Resources Tab online. This may be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.

          Target Information

          RELM alpha, also known as Resistin-like alpha or FIZZ1, is a small cytokine (88 aminoacids, MW 9.4 kDa) produced mostly by macrophages. Its highest constitutive expression has been observed in small peritoneal macrophages, alveolar macrophages and adipose tissue macrophages. It is strongly induced by IL-4 and IL-13. Together with Arginase 1 and Ym-1, RELM alpha is considered one of the hallmarks of the M2 polarization of mouse macrophages, however, macrophages polarized with IL-10 (M2c) express neither RELM alpha nor Arginase 1. Also, the expression patterns of RELM alpha and Arginase 1 in untreated tissue macrophages significantly differ. Elevated levels of RELM alpha may indicate parasitic infection. Humans do not have RELM alpha gene but human Resistin resembles mouse RELM alpha in respect to sequence, tissue distribution and function. Other members of RELM family in mouse: RELM beta, RELM gamma and Resistin display different tissue distribution and functional properties. RELM alpha plays an important role in parasite infection, mucosal inflammation and allergy.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: cysteine-rich secreted protein A12-gamma; cysteine-rich secreted protein FIZZ1; FIZZ2; found in inflammatory zone 1; HXCP2; hypoxia-induced mitogenic factor; parasite-induced macrophage novel gene 1 protein; PMNG1; FIZZ1; RELM-beta; RELMalpha; FIZZ1; RELMb; RELMbeta; similar to cysteine-rich secreted A12-alpha protein; unnamed protein product

          View more View less

          Gene Aliases: 1810019L16Rik; Fizz-1; Fizz1; HIMF; RELM-alpha; RELMa; RELMalpha; Xcp2

          View more View less

          UniProt ID: (Mouse) Q9EP95

          View more View less

          Entrez Gene ID: (Mouse) 57262

          View more View less

          Function(s)
          hormone activity
          Process(es)
          signal transduction response to nematode response to selenium ion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-5d6d8bc979-gw77j:80/100.66.76.64:80.
          git-commit: e71199a02a6c1449b2ac9ded8316d5734e8aa2de
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.62.0-2026.11.45-1.0