Nalgene™ Polypropylene Griffin Low-Form Plastic Beakers
Nalgene™ Polypropylene Griffin Low-Form Plastic Beakers
Nalgene™ Polypropylene Griffin Low-Form Plastic Beakers
Nalgene™ Polypropylene Griffin Low-Form Plastic Beakers
Nalgene™ Polypropylene Griffin Low-Form Plastic Beakers
Thermo Scientific™

Nalgene™ Polypropylene Griffin Low-Form Plastic Beakers

Carry out precise and efficient pouring with these low-form beakers, featuring a single-spout design and easy-to-read silk-screened graduations.
Catalog NumberCapacity (Metric)
1201-0050PK
also known as 1201-0050
50 mL
1201-0030PK
also known as 1201-0030
30 mL
1201-0100PK
also known as 1201-0100
100 mL
1201-0150PK
also known as 1201-0150
150 mL
1201-0250PK250 mL
1201-0400PK
also known as 1201-0400
400 mL
1201-0600PK600 mL
1201-1000PK1000 mL
1201-2000PK
also known as 1201-2000
2000 mL
1201-4000PK4000 mL
Catalog number 1201-0050PK
also known as 1201-0050
Price (JPY)
4,020
Online offer
Ends: 25-Sep-2026
6,720
Save 2,700 (40%)
Pack of 12
Capacity (Metric):
50 mL

Carry out precise and efficient pouring with Thermo Scientific™ Nalgene™ Polypropylene Griffin Low-Form Beakers, featuring a single-spout design and easy-to-read silk-screened graduations. Rugged, reusable, construction that will last for years under typical lab conditions. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables. Meets ISO/DIN 7056 International Standards for Laboratory Plasticware Accuracy. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.

Features:

  • Ribs for easy stacking
  • Silk-screened with graduations, size code, resin code, catalog number, max. temperature and No Flame symbol for easy identification and safe use
  • Autoclavable/Graduated
  • Meets ISO/DIN 7056 International Standards for Laboratory Plasticware Accuracy
  • Participates in My Green Lab's ACT Label program for Accountability, Consistency and Transparency (ACT) around evaluating product sustainability

Ordering Information: Available in a size variety pack using Cat. No. 1201-1234

Recycling Information: Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Thermo Fisher Scientific encourages recycling Nalgene polypropylene labware at end of product life in communities where polypropylene can be recycled.

Nalgene beakers and graduated cylinders are delivered in corrugate cartons made from 100% post-consumer recycled material. Packaging bags are made from LDPE and are readily recyclable once the label is removed. Please continue the cycle and recycle your cartons and bags.

Specifications
ACT LabelYes
ColorNatural translucent
DescriptionBeaker Griffin Low-Form PP 50 mL
Graduation Range10 to 50 mL
Increments5 mL
Quantity12/Pk.; 48/Cs.
TypeBeaker
AutoclavableAutoclavable
Capacity (Metric)50 mL
Certifications/ComplianceISO/DIN 7056
DisposableReusable
GraduatedYes
MaterialPPCO
Scale10mL to 50mL in 5mL increments
StyleGriffin, Low Form
Unit SizePack of 12

Frequently asked questions (FAQs)

Several of your labware products are available in polypropylene (PP, PPCO) or polymethylpentene (PMP). Which should I choose?

These two materials were chosen as labware raw materials for their durability, broad chemical compatibility profile, and wide range of use temperature. Polymethylpentene offers a higher level of transparency (comparable to glass) but is slightly more brittle and therefore somewhat more likely than polypropylene to crack if dropped. Chemical or environmental factors in some applications may strongly indicate the use of only one material- please refer to laboratory plastics reference materials or call Thermo Scientific Laboratory Products Essentials technical support for assistance.