PureLink™ RNase A (20 mg/mL)
Invitrogen™

PureLink™ RNase A (20 mg/mL)

PureLink™ RNase A (ribonuclease A) is a bovine pancreatic ribonuclease that cleaves single-stranded RNA. This reagent is also a componentRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
1209103925 mL
1209102110 mL
Catalog number 12091039
Price (CLP)
555.968
Each
Quantity:
25 mL
Price (CLP)
555.968
Each

PureLink™ RNase A (ribonuclease A) is a bovine pancreatic ribonuclease that cleaves single-stranded RNA. This reagent is also a component of the PureLink™ plasmid purification system.

Concentration
20 mg RNase A/mL in 50 mM Tris-HCl (pH 8.0), 10 mM EDTA.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Packaging TypeBottle
Product LineUltraPure
Quantity25 mL
Shipping ConditionRoom Temperature
Concentration20 mg/mL
FormLiquid
Unit SizeEach
Contents & Storage
1 x 25 mL bottle.
Store at 2–8°C.

Frequently asked questions (FAQs)

I need to digest my total RNA for my ribonuclease protection assay. What RNase should I use?

Total RNA can be digested using RNAse A or RNase T1. A mix of both can be used in ribonuclease protection assays.