4-Hydroxyquinoline-2-carboxylic acid, hydrate, 98%
4-Hydroxyquinoline-2-carboxylic acid, hydrate, 98%
4-Hydroxyquinoline-2-carboxylic acid, hydrate, 98%
Thermo Scientific Chemicals

4-Hydroxyquinoline-2-carboxylic acid, hydrate, 98%

CAS: 345909-35-5 | C10H7NO3 | 189.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 121910050
Price (USD)
115.65
Online Exclusive
128.00
Save 12.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
115.65
Online Exclusive
128.00
Save 12.35 (10%)
Each
Chemical Identifiers
CAS345909-35-5
IUPAC Name4-hydroxyquinoline-2-carboxylic acid
Molecular FormulaC10H7NO3
InChI KeyHCZHHEIFKROPDY-UHFFFAOYSA-N
SMILESOC(=O)C1=NC2=CC=CC=C2C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to brown
Appearance (Form)Powder
Infrared spectrumConforms
Titration excess NaOH97.5 to 102.5 % (On dry substance)
Water7 to 10 % (K.F.)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only