Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MDIKQMKNF VSQELKGLKQ EHRLLSLHIG ACESIMKKKT KQDFQELIKT EHALLEGFNI RESTSYIEEH IDRQVSPIES LRLMCLLSIT ENGLIPKDYR SLKTQYLQSY GPEHLLTFSN LRRAGLLTEQ APGDTLTAVE SKVSKLVTDK AAGKITDAFS SLAKRSNFRA ISKKLNLIPR VDGEYDLKVP RDMAYVFSGA YVPLSCRIIE QVLERRSWQG LDEVVRLLNC SDFAFTDMTK EDKASSESLR LILVVFLGGC TFSEISALRF LGREKGYRFI FLTTAVTNSA RLMEAMSEVK A (318-617 aa encoded by BC016445)
Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene is a member of the Sec-1 domain family, and encodes the human ortholog of rat Vps33b which is homologous to the yeast class C Vps33 protein. The mammalian class C Vps proteins are predominantly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ14848; hVPS33B; r-vps33b; unnamed protein product; vacuolar protein sorting 33 homolog B; vacuolar protein sorting 33 homolog B (yeast) isoform VPS33B_1; vacuolar protein sorting 33 homolog B (yeast) isoform VPS33B_2; vacuolar protein sorting 33 homolog B (yeast) isoform VPS33B_3; vacuolar protein sorting 33-like protein B; vacuolar protein sorting 33B; vacuolar protein sorting homolog r-vps33b; Vacuolar protein sorting-associated protein 33B; VPS33B
Gene Aliases: KDIDAR; PFIC12; VPS33B
UniProt ID: (Human) Q9H267, (Mouse) P59016, (Rat) Q63616
Entrez Gene ID: (Human) 26276, (Mouse) 233405, (Rat) 64060
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support