Indole, 99+%
Indole, 99+%
Indole, 99+%
Thermo Scientific Chemicals

Indole, 99+%

CAS: 120-72-9 | C8H7N | 117.15 g/mol
Quantity:
10 g
25 g
100 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 122150025
Price (JPY)
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS120-72-9
IUPAC Name1H-indole
Molecular FormulaC8H7N
InChI KeySIKJAQJRHWYJAI-UHFFFAOYSA-N
SMILESN1C=CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point51°C to 53°C
Appearance (Color)White to light pink
Infrared spectrumConforms
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only